Mouse Recombinant Growth hormone (from E. coli)
Supplier: BioVision
4770-1000 4770-50
10007-132EA 4261.35 USD
10007-132 10007-134
Mouse Recombinant Growth hormone (from E. coli)
>95% Pure active Recombinant murine Growth Hormone.
Recombinant mouse Growth Hormone is fully biologically active when compared to standard human growth hormone which is 3 units/mg.
GH is a member of the somatotropin/prolactin family of hormone which plays an important role in growth control. Recombinant mouse Growth Hormone produced in E.Coli is a single, non-glycosylated, single chain containing 191 amino acids and having a molecular mass of 22.0 kDa.