Would you like to log in again?
EPO-alpha, human recombinant, BioVision
Supplier: BioVision
4764-500
10007-122EA 252.35 USD InStock
10007-122
EPO-alpha, human recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant human EPO-alpha.
BioVision's Erythropoietin is fully biologically active when compared to standard. The Specific Activity measured by Normocytic anemic mice was found to be 120,000IU/mg.
Recombinant Human Erythropoietin-α produced in Chinese hamster ovary (CHO) cells by recombinant DNA technology is a single, polypeptide chain containing 165 amino acids and having a predicted molecular mass of 21 kDa and apparent glycosylated molecular mass of 30.4 kDa. EPO-α is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty