Would you like to log in again?
Industrial Balances, Standard, CKW Series, Ohaus®
Supplier: OHAUS SCALE CORP
80251044 80251042 80251043
87004-222EA 1984.25 USD InStock
87004-222 87004-218 87004-220
Industrial Balances, Standard, CKW Series, Ohaus®
Balances and Scales
Advanced checkweighing bench scale.
  • An IP 67 stainless steel load cell and IP 66 stainless steel indicator make this range particularly suited to static checkweighing in industrial and food processing environments
  • Easy set up
  • Quick stabilisation of under 2 seconds speeds the weighing process
  • Adjustable LED display with configurable features e.g. flashing result, bar graph and audible alarm
  • Designed for wash down of both base and indicator
The base has open architecture framework that is designed for cleaning in wash down environments and the base also has rounded corners and a removable platform with an external skirt. The checkweighing operation has the following features; fast target entry to expedite the input of set-point values and the menus allow different modes of operation including positive or negative checkweighing. In addition the balance features a 20-part Library storage for Item ID no., Over, Under, Target and Pre-set Tare values. The display has been designed to help the operator attain the target weight with visual and audible signals to indicate under or overweight.
Certifications: Supports HACCP compliant.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty