Would you like to log in again?
Endostatin, human recombinant, BioVision
Supplier: BioVision
4759-1000 4759-100 4759-20
10007-112EA 1133 USD InStock
10007-112 10007-110 10007-114
Endostatin, human recombinant, BioVision
Proteins and Peptides
>95% Pure Recombinant human Endostatin.
Endostatin has been identified as a C-terminal fragment of Collagen type 18, a recently identified member of a family of collagen-like proteins referred to as multiplexin family Endostatin specifically inhibits proliferation of endothelial cells although it does not affect the proliferation of EOMA cells. Endostatin also potently inhibits angiogenesis and tumor growth. Endostatin has an important role in endothelial cell adhesion and cytoskeletal organization. Endostatin can be found in vessel walls (elastic fibers) and basement membranes. Recombinant human Endosatin expressed in yeast causes G1 arrest of endothelial cells, and endostatin treatment results in apoptosis of HUVE and HMVE cells.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty