Would you like to log in again?
TFF-3, human recombinant, BioVision
Supplier: BioVision
4746-100 4746-1000 4746-20
10006-954EA 612.85 USD InStock
10006-954 10007-100 10007-102
TFF-3, human recombinant, BioVision
Proteins and Peptides
>97% Pure active Recombinant human TFF-3.
Determined by its ability to chemoattract human MCF-7 cells using a concentration 1.0-10.0 ng/ml.
Proteins of the TFF family are characterized by having at least 1 copy of the trefoil motif, a 40-amino acid domain that contains 3 conserved disulfides. Trefoil Factors are stable secretory proteins expressed in gastrointestinal mucosa which protect the mucosa from insults, stabilize the mucus layer and affect healing of the epithelium. TFF3 induces ciliogenesis and promotes airway epithelial ciliated cell differentiation, through an EGFR-dependent pathway.TFF-3 is normally expressed in hepatocellular carcinoma and its overexpression is crucial for progression in mouse and human hepatocellular carcinogenesis. TFF-3 Human Recombinant produced in E.Coli is a homodimeric, non-glycosylated, polypeptide chain containing 2 x 59 amino acid chains which includes a 40 amino acid trefoil motif containing 3 conserved disulfide bonds and has MW = 13.2 kDa. TFF-3 Human Recombinant is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty