CellExp™ Human Recombinant Progranulin (from HEK293 Cells)
Supplier: BioVision
CellExp™
AB113403-10UG
10006-950EA 574.57 USD
10006-950
CellExp™ Human Recombinant Progranulin (from HEK293 Cells)
Proteins and Peptides
>90% Pure Recombinant human Progranulin.
Measured by chemotaxis using THP-1 cells.
Progranulin, also called proepithelin and PC cell-derived growth factor, is a single precursor protein of granulins which are a family of secreted, glycosylated peptides that are cleaved from a single precursor protein with 7.5 repeats of a highly conserved 12-cysteine granulin/epithelin motif. Granulins are a variety of active, 6.0 kDa peptides and named granulin A (epithelin 1), granulin B (epithelin 2), granulin C, etc. Both the peptides and intact progranulin protein regulate cell growth. However, different members of the granulin protein family may act as inhibitors, stimulators, or have dual actions on cell growth. Granulin family members are important in normal development, wound healing, and tumorigenesis. Progranulin Human Recombinant fused to FLAG at C-terminus produced in HEK is a single, glycosylated, polypeptide chain containing 593 amino acids and having a molecular mass of 88.0 kDa. The Progranulin is purified by standard chromatographic techniques.