Would you like to log in again?
Human CellExp™ Progranulin, Rat Recombinant, BioVision
Supplier: BioVision
4735-50 4735-10
10006-940 10006-834
Human CellExp™ Progranulin, Rat Recombinant, BioVision
>95% Pure Recombinant rat Progranulin.
Progranulin (PGRN), also called proepithelin and PC cell-derived growth factor, is a single precursor protein of granulins which are a family of secreted, glycosylated peptides. It is a widely expressed pluripotent growth factor which plays a role in processes such as development, wound repair and inflammation by activating signaling cascades that control cell cycle progression and cell motility. Its function in the central nervous system is of interest, as mutations in the PGRN gene were found in cases of fronto-temporal degeneration (FTLD). In addition, PGRN has also been linked to tumorigenesis. Progranulin is a biomarker for FTLD, other types of Alzheimer‘s Disease (AD) and potentially for MCI (Mild Cognitive Impairment). Additionally, PGRN is described as a new ligand of TNF receptors and a potential therapeutic against inflammatory disease like arthritis. Rat Recombinant Progranulin fused to FLAG® at C-terminus produced in HEK is a single, glycosylated, polypeptide chain purified by standard chromatographic techniques, containing 602 amino acids and having a molecular mass of ~65.0 kDa.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty