Would you like to log in again?
Heregulin-β2 (HRG2β), Human Recombinant , BioVision
Supplier: BioVision
4730-1000 4730-10 4730-50
10007-082 10007-080 10007-084
Heregulin-β2 (HRG2β), Human Recombinant , BioVision
>95% Pure active Recombinant human Heregulin-β2 (HRG2β)
Neuregulin/Heregulin is a family of structurally related polypeptide growth factors derived from alternatively spliced genes (NRG1, NRG2, NRG3 and NRG4). To date, there are over 14 soluble and transmembrane proteins derived from the NRG1 gene. Proteolytic processing of the extracellular domain of the transmembrane NRG1 isoforms release soluble growth factors. HRG-β2 is a signaling protein for ErbB2/ErbB4 receptor on the cardiac muscle cells. The polypeptide is comprised of an immunoglobulin and epidermal growth factor-like (EGF-like) domains.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty