Would you like to log in again?
C5a (His-Tag), human recombinant, BioVision
Supplier: BioVision
4995A-1000 4995A-20 4995A-100
10007-398EA 5098.5 USD InStock
10007-398 10007-400 10007-396
C5a (His-Tag), human recombinant, BioVision
Proteins and Peptides
>95% Pure active Recombinant Human C5a (His-Tag)
Human recombinant C5a is as active as that of serum derived C5a based on enzyme release assay of myeloperoxidase. The current lot of ED₅₀ = 1.36 nmolar.
Human complement C5a is a 74-amino acid glycopeptide generated during complement activation from the a-chain of complement C5. C5a is a potent chemotactic factor for human peripheral blood neutrophils and monocytes, and is believed to play an important role in a number of inflammatory conditions. The 74 amino acid, human C5a protein expressed in E. coli with a 6 amino acid, N-terminal His·Tag sequence and 28 amino acid proprietary vector sequence necessary for expression (C5a without His-tag is alailable, BioVision Cat # 4995B). The recombinant protein is not further processed to remove the additional sequence. Recombinant C5a has an activity similar to that of serum derived C5a based on enzyme assay of myeloperoxidase release.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty