Would you like to log in again?
Activin A, human recombinant, BioVision
Supplier: BioVision
4724-10 4724-100 4724-1000
10007-062 10007-064 10007-066
Activin A, human recombinant, BioVision
>95% Pure active Recombinant human Activin A.
The activity is determined by the ability to induce cytoxicity of MPC-11 cells and is typically less than 2 ng/mL.
Activin A is a member of the TGFbeta family of proteins produced by many cell types throughout development. Activins interact with Type I and Type II serine/threonine kinases to signal to SMAD proteins to regulate a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, metabolism, etc. Activin A is a homodimer of two beta A chains and is not biologically active until the N terminal propeptide is cleaved from each. Human Activin A has 100% amino acid sequence identity to mouse, rat, porcine, bovine and feline proteins. Recombinant human Activin A is a non-glycosylated homodimer, containing two 117 amino acid chains, with a total molecular weight of 26.2 kDa.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty