Would you like to log in again?
PARP-1, human recombinant, BioVision
Supplier: BioVision
4992-50
10007-388EA 376.99 USD InStock
10007-388
PARP-1, human recombinant, BioVision
Proteins and Peptides
>95% Pure active Recombinant human PARP-1.
1000 U/vial, specific activity = 20000 U/mg PARP1. 1U=10 fmol ADP-ribose incorporated into 5 μg immobilized histone in 30 min at room temperature. Note: Activity measurements are approximate values.
Poly-ADP-ribose metabolism plays a major role in a wide range of biological processes, such as maintenance of genomic stability, transcriptional regulation, energy metabolism and apoptosis. PARP1 Enzyme, is highly purified and enzymatically active human PARP1, expressed in a baculovirus expression system. Upon addition of NAD+ and cofactors the PARP1 is automodified by the addition of ADP-ribose monomers to form poly(ADP-ribose) polymer. The PARP1 is supplied lyophilized, and upon reconstitution with water, it retains its native poly ADP-ribosylation enzymatic activity. It is useful for high-throughput enzymatic assays, visualization of the automodification reaction by SDS-PAGE and Western blotting, ELISA, standard for SDS-PAGE, and in other assays.
Spin tube in a microfuge for 15 sec to sediment lyophilized material. Carefully open the vial and add 100 μL dH₂O. Vortex gently for 20 sec (avoid air bubbles). Let stand for 5 min. Carefully triturate the sample 10-times using a pipetman (avoid air bubbles). Spin briefly in microfuge to consolidate. The final concentrations are as follows 20 mm Tris, pH 8, 0.3 M NaCl, 0.1 mm EDTA, 1 mM DTT, plus lyophilization stabilizers.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty