Would you like to log in again?
IGF-BP5, human recombinant, BioVision
Supplier: BioVision
4723-25 4723-100 4723-1000
10007-060EA 195.7 USD InStock
10007-060 10007-056 10007-058
IGF-BP5, human recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant human IGF-BP5.
The ED₅₀ as determined by its ability to inhibit IGF-II induced proliferation of MCF-7 is <0.3 mg/ml, in the presence of 15 ng/ml of human IGF-II.
Human IGF-BP5 is a 28.6 kDa cysteine-rich secreted protein produced by Vascular smooth muscle cells. It is the major IGF-binding protein present in bone tissue and helps potentiate the action of IGF-I on smooth muscle cells, fibroblasts or osteoblasts. Studies show that IGF-BP5 acts as a growth inhibitor and pro-apoptotic agent in breast cancer cells. IGF-BP5 overexpressing mice show an increase in neonatal mortality, reduced female fertility, whole body growth inhibition, and retarded muscle development. Recombinant human IGF-BP5 is a 28.6 kDa protein consisting of 253 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty