Would you like to log in again?
IGF-BP3, human recombinant, BioVision
Supplier: BioVision
4720-100 4720-25 4720-1000
10007-050EA 540.75 USD InStock
10007-050 10007-054 10007-052
IGF-BP3, human recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant human IGF-BP3.
The ED₅₀ as determined by its ability to inhibit IGF-II induced proliferation of MCF-7 is <0.2 mg/ml, in the presence of 15 ng/ml of human IGF-II.
Human IGF-BP3, also known as growth-hormone-dependant binding protein, is a 28.9 kDa cysteine-rich secreted protein. It is the major IGF binding protein present in the plasma of human and animals. In addition to its ability to modulate the activity of IGF-I and IGF-2, IGF-BP3 also exerts inhibitory effects on follicle stimulating hormone (FSH) activity. Decreased plasma levels of IGF-BP3 often results in dwarfism, whereas elevated levels of IGF-BP3 may lead to acromegaly. The expression of IGF-BP3 in fibroblasts is stimulated by mitogenic growth factors such as Bombesin, Vasopressin, PDGF, and EGF. Recombinant human IGF-BP3 is a 28.8 kDa protein consisting of 264 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty