Would you like to log in again?
Human CellExp™ IL-12, Mouse Recombinant, BioVision
Supplier: BioVision
4719-10 4719-100
10007-046 10007-048
Human CellExp™ IL-12, Mouse Recombinant, BioVision
>95% Pure active Recombinant murine IL-12.
The activity is determined by the induced expression of CD25 on CD8+ mouse T cells and is typically detectable starting at 0.1 ng/mL.
Interleukin-12 (IL-12) is an important mediator of cellular-immunity produced by dendritic cells, macrophages and B cells when stimulated with antigen. IL-12 is involved in differentiating precursor helper T cells to Th1 and/or Th2 subtypes and promotes the production of IFN gamma and TNF alpha. Mouse IL-12 is active on human and mouse cells. Recombinant mouse IL-12 is a glycosylated, disulfide-linked heterodimer. It is containing one p35 subunit (35 kDa) and one p40 subunit (40 kDa), with a total of 506 amino acids and a molecular weight of approximately 70 kDa.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty