Would you like to log in again?
Follistatin, mouse recombinant, BioVision
Supplier: BioVision
4718-20 4718-100 4718-1000
10007-044EA 267.72 USD InStock
10007-044 10007-040 10007-042
Follistatin, mouse recombinant, BioVision
Proteins and Peptides
>90% Pure active Recombinant murine Follistatin.
The activity is determined by the dose-dependent neutralization of 7.5 ng/mL human Activin A. Complete neutralization is typically reached at less than 0.3 ug/mL.
Follistatin is an autocrine acting protein that is expressed by many tissues, but at noteably higher levels in the ovary and skin. Follistatin functions to negatively regulate the signaling of a wide variety of TGF-beta family members (activin, BMPs, myostatin, GDF-11 and TGF-beta 1). Mechanistically, follistatin works as an antagonist by complexing with TGF-beta family members to prevent them from interacting with their signaling receptors. Recombinant mouse follistatin is a nonglycosylated protein, containing 289 amino acids, with a molecular weight of 31.6 kDa.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty