Would you like to log in again?
Neuregulin/Heregulin-1β (NRG-1β/HRG-1β), human recombinant , BioVision
Supplier: BioVision
4711-10 4711-1000 4711-50
10007-022EA 87.55 USD InStock
10007-022 10007-024 10007-026
Neuregulin/Heregulin-1β (NRG-1β/HRG-1β), human recombinant , BioVision
Proteins and Peptides
>98% Pure Recombinant human Heregulin beta-1.
The ED₅₀ was determined by the dose-dependent stimulation of the proliferation of human MCF-7 cells is < 0.5 ng/ml, corresponding to a specific activity of > 2 x 10⁶ units/mg.
Neuregulin/Heregulin is a family of structurally related polypeptide growth factors derived from alternatively spliced genes (NRG1, NRG2, NRG3 and NRG4). To date, there are over 14 soluble and transmembrane proteins derived from the NRG1 gene. Proteolytic processing of the extraCellular domain of the transmembrane NRG1 isoforms release soluble growth factors. HRG1-β1 contains an Ig domain and an EGF-like domain that is necessary for direct binding to receptor tyrosine kinases erb3 and erb4. This binding induces erb3 and erb4 heterodimerization with erb2, stimulating intrinsic kinase activity, which leads to tyrosine phosphorylation. Although HRG1-β1 biological effects is still unclear, it has been found to promote motility and invasiveness of breast cancer cells which may also involve up-regulation of expression and function of the autocrine motility-promoting factor (AMF). Recombinant human Heregulin-β1 (HRG1-β1) is a 7.5 kDa polypeptide consisting of only the EGF domain of heregulin-β1 (65 amino acid residues).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty