Would you like to log in again?
Heat Shock Protein 90 (Hsp90 beta) NT-His Tag, human recombinant, BioVision
Supplier: BioVision
4858H-1000 4858H-25 4858H-100
10007-224 10007-226 10007-222
Heat Shock Protein 90 (Hsp90 beta) NT-His Tag, human recombinant, BioVision
>99% Pure Recombinant human Heat Shock Protein 90.
Not available at this time
Hsp90 is a molecular chaperone and is one of the most abundant proteins in unstressed cells. In mammalian cells, there are two genes encoding cytosolic Hsp90 homologues, with the human Hsp90α showing 85% sequence identity to Hsp90β. The recombinant Hsp90β produced from Sf9 cells represents the full length (a.a. 1-723) of Hsp90β subunit and a N-term His-tag.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty