Mouse Recombinant RELM beta (from E. coli)
Supplier: BioVision
4655-100 4655-25 4655-1000
10006-822EA 951.51 USD
10006-822 10006-826 10006-824
Mouse Recombinant RELM beta (from E. coli)
>97% Pure Recombinant murine RELM-beta.
RELM-beta (Resistin-Like Molecule-beta) is a member of a recently identified family of secreted proteins containing a conserved cystein-rich C-terminus. The RELM family consists of resistin (also called FIZZ3), RELM-alfa (FIZZ1), RELM-beta (FIZZ2) and RELM-gamma. Only resisistin and RELM-beta were found in humans whereas all four RELM family members were identified in rodents. Recombinant Human Relm-beta is a non-glycosylated polypeptide chain consisting of 83 amino acids with a molecular weight of 8,893 kDa.