Would you like to log in again?
RELM-beta, human recombinant, BioVision
Supplier: BioVision
4654-25 4654-100 4654-1000
10006-820EA 185.4 USD InStock
10006-820 10006-928 10006-930
RELM-beta, human recombinant, BioVision
Proteins and Peptides
>95% Pure Recombinant human RELM-beta.
Test in process
Human RELM β (Resistin-like molecule β/FIZZ2) is a new member to the family of adipocyte secreted proteins called adipocytokines. This family includes the RELM α, RELM β and Resistin molecules. Interestingly, RELM β and Resistin share similar characteristics such as an additional cysteine residue within the variable N-terminal region and are both homodimeric proteins. However, the RELM β is expressed only in the gastrointestinal track; especially the colon, while the Resistin and RELM β are secreted exclusively by adipocytes. Currently, the biological function of these proteins, as well as their molecular targets is largely unknown. Recombinant Human RELM β is a disulfide-linked homodimer with a total molecular weight of 11.0 kDa, consisting of 90 amino acid residue chains.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty