Would you like to log in again?
UbcH5c, human recombinant (His-tag), BioVision
Supplier: BioVision
4854-100
10007-220EA 360.5 USD InStock
10007-220
UbcH5c, human recombinant (His-tag), BioVision
Proteins and Peptides
>95% Pure active Recombinant human UbcH5c.
UbcH5c is highly homologous to the S. cerevisiae UbC4/5 family of E2 enzymes, demonstrated to be essential for degradation of regulatory and abnormal proteins. UbcH5c, together with UbcH5a and UbcH5b, are the most active class of E2 enzymes in cell extracts and are associated with regulation of a number of transcription factors including p53. The His-tagged version of this enzyme is not susceptible to self-ubiquitinylation, which can occur with GST-tagged versions. Typical enzyme concentration to support in vitro conjugation is 100 nM to 1 µM depending upon the conditions.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty