Would you like to log in again?
PTH, human recombinant, BioVision
Supplier: BioVision
4650-50 4650-1000
10006-926 10006-924
PTH, human recombinant, BioVision
>98% Pure active Recombinant human PTH.
Recombinant human PTH is fully active when compared to standards. The activity calculated by μmR10⁶ cells/cAMP method corresponds to a specific activity of 1 x 10⁴ IU/mg.
Parathyroid Hormone (PTH) is the most important endocrine regulator of calcium and phosphorus concentration in extraCellular fluid. This hormone is secreted from cells of the parathyroid glands and targets cells in bone and kidney. Like other protein hormones, parathyroid hormone is synthesized as a preprohormone. After intraCellular processing, the mature hormone is packaged within the Golgi into secretary vesicles and secreted into blood by exocytosis. Recombinant human PTH is a 9 kDa protein consisting of 84 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty