Sample Cylinder 316 SS Hollow Hex Plug, Restek
Supplier: Restek
26377
10145-946EA 22.84 USD
10145-946
Sample Cylinder 316 SS Hollow Hex Plug, Restek
Cylinders Hydrometer Cylinders
Sample cylinder plugs protect Swagelok tube fitting or NPT end connections on valves during cylinder transport.
  • Stainless steel fabrication
Fitting type: 1/4" male NPT