Closed End SPE Cartridge: Activated Sodium Sulfate, Restek
Supplier: Restek
26207
10059-522PK 476.43 USD
10059-522
Closed End SPE Cartridge: Activated Sodium Sulfate, Restek
Chromatography Columns
Endcapped SPE cartridges of activated sodium sulfate are useful in removing excess water from solvent solutions prior to analyses.
  • High quality anhydrous sodium sulfate
  • Approximately 2 grams prepackaged in a convenient capped cartridge with both male and female luer ends for easy connection to a variety of devices or equipment
  • Capped cartridges will remain active for long periods of storage in the lab
The adsorbent is fully activated and ready to use for removal of excess water from organic solvent solutions prior to many types of analysis.