Anti-MLLT2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP6189C
60942-640EA 751.67 USD
60942-640
Anti-MLLT2 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: MLLT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR