Human Recombinant Heat Shock 27 (from E. coli)
Supplier: BioVision
4853-1000 4853-50 4853-10
10007-214EA 385.27 USD
10007-214 10007-216 10007-218
Human Recombinant Heat Shock 27 (from E. coli)
>98% Pure active Recombinant human heat shock protein 27.
Immunoreactivity is confirmed by reaction with monoclonal mouse antibodies against HSP-27.
Recombinant Human Heat Shock Protein produced in E.Coli is a single, non-glycosylated polypeptide chain containing 205 amino acids and have a molecular weight of 22.7 kDa. The protein was purified by proprietary chromatographic techniques.