Would you like to log in again?
UbcH5a, human recombinant (His-tag) (New size), BioVision
Supplier: BioVision
4851-10 4851-100
10007-206 10007-208
UbcH5a, human recombinant (His-tag) (New size), BioVision
>95% Pure active Recombinant human UbcH5a.
UbcH5a is highly homologous to the S. cerevisiae Ubc4/5 family of E2 enzymes demonstrated to be essential for degradation of regulatory and abnormal proteins. UbcH5a, together with UbcH5b and UbcH5c, are the most active class of E2 enzymes in cell extracts and are associated with regulation of a number of transcription factors. Specifically, UbcH5a stimulates the conjugation of ubiquitin to the tumor suppressor p532. Useful for in vitro ubiquitinylation reactions. Typical enzyme concentration to support conjugation in vitro is 100 nM to 1 µM depending upon conditions.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty