Would you like to log in again?
Cloud Chamber with source
Supplier: SCIENCE FIRST INC SE
16760
470205-904EA 139 USD InStock
470205-904
Cloud Chamber with source
Educational Materials Earth Science Educational Materials Meteorology Learning Activities
This reusable activity illustrates earth’s rotation and revolution.
  • Kit Introduces Concepts of Radioactive Decay
  • Students Track the Movement of Alpha and Beta Particles
  • Reusable
It also introduces concepts of radioactive decay. With this detector, the tracks of radioactive particles are made visible by the condensation of droplets in a thin layer of alcohol vapor.
It includes a mail-away coupon for the source of alpha and beta particles and requires liquid nitrogen or dry ice.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty