Would you like to log in again?
UbcH2, human recombinant (His-tag), BioVision
Supplier: BioVision
4848-100
10007-202EA 360.5 USD InStock
10007-202
UbcH2, human recombinant (His-tag), BioVision
Proteins and Peptides
>95% Pure active Recombinant human UbcH2.
UbcH2 is the human homolog to the yeast DNA repair gene RAD6, which is induced by DNA damaging reagents. It has been shown to conjugate ubiquitin to histone H2A in an E3 dependent manner in vitro. UbcH2 consists of two functionally independent domains, an N-terminal core domain with ubiquitin conjugating activity, and a C-terminal domain that interacts with substrate proteins. This protein is useful for in vitro ubiquitinylation reactions. The His-tagged version of this enzyme is not susceptible to self-ubiquitinylation, which can occur with GST-tagged versions. Typical enzyme concentration to support conjugation in
vitro is 100 nM to 1 μM depending upon conditions.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty