Would you like to log in again?
Galectin-1, human recombinant, BioVision
Supplier: BioVision
4648-10 4648-1000 4648-50
10006-918EA 87.55 USD InStock
10006-918 10006-920 10006-922
Galectin-1, human recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant human Galectin-1.
Determined by its ability to chemoattract human blood monocytes. Chemotactic activity was observed at a concentration of 2.5 µg/ml with a peak response obtained at 250 µg/ml.
Galectin-1 belongs to growing family of evolutionary conserved animal lectins. Galectins consists of β-galactoside binding lectins that contain homologous carbohydrate recognition domains (CRDs). Galectin-1 has the ability to induce apoptosis of activated T-cells and T-leukaemia cell lines. Other activities include cell differentiation and inhibition activity of CD45 protein phosphatase activity. Galectin also binds β-galactoside as well as CD45, CD3 and CD4. Recombinant Human Galectin is a 14.5 kDa protein consisting of 134 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty