Human Recombinant VEGFD (from E. coli)
Supplier: BioVision
4643-50 4643-1000 4643-10
10006-910EA 694.66 USD
10006-910 10006-908 10006-906
Human Recombinant VEGFD (from E. coli)
>98% Pure active Recombinant human VEGF-D.
VEGF-D is most closely related to VEGF-C by virtue of the presence of N- and C-terminal extensions that are not found in other VEGF family members. In adult human tissues, VEGF-D mRNA is most abundant in heart, lung, skeletal muscle, colon, and small intestine. Analyses of VEGF-D receptor specificity revealed that VEGF-D is a ligand for both VEGF receptors (VEGFRs) VEGFR-2 (Flk1) and VEGFR-3 (Flt4) and can activate these receptors. However, VEGF-D does not bind to VEGFR-1. Expression of a truncated derivative of VEGF-D demonstrated that the receptor-binding capacities reside in the portion of the molecule that is most closely related in primary structure to other VEGF family members and that corresponds to the mature form of VEGF-C. In addition, VEGF-D is a mitogen for endothelial cells. Recombinant VEGF-D was expressed based on sequence from Genebank Accession# D89630, 20-354 aa.