Would you like to log in again?
VEGF-C, rat recombinant, BioVision
Supplier: BioVision
4635-10 4635-50 4635-1000
10006-900EA 213.09 USD InStock
10006-900 10006-904 10006-902
VEGF-C, rat recombinant, BioVision
Proteins and Peptides
>90% Pure active Recombinant rat VEGF-C.
Testing in progress
VEGF-C (Vascular Endothelial Growth Factor-C) is a 125 amino acid recombinant protein that lacks the N-terminal and C-terminal extensions and contains only the middle third VEGF homology domain.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty