Would you like to log in again?
sTNF-RI, human recombinant, BioVision
Supplier: BioVision
4348-1000 4348-20 4348-100
10006-586EA 2987 USD InStock
10006-586 10006-588 10006-584
sTNF-RI, human recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant human sTNF-RI.
The biological activity of sTNF-RI was determined by its inhibitory effect of the TNF-α mediated cytotoxicity in murine L-929 cells. The ED₅₀ for the effect in the presence of 0.25 ng/ml of recombinant TNF-α, is 0.05 mg/ml.
Tumor Necrosis Factor receptor type I (TNF-RI) is also referred as p60. The p60 form of the receptor is primarily expressed on epithelial cells. Recombinant Human soluble TNF receptor Type I (sTNF-RI) is a 18.3 kDa protein containing 162 amino acid residues comprising the extra Cellular domain of TNF Receptor Type I.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty