Human Recombinant UBCH1 (from E. coli), GST Tag
Supplier: BioVision
AB111465-100UG
10007-200EA 1087.87 USD
10007-200
Human Recombinant UBCH1 (from E. coli), GST Tag
Proteins and Peptides
>95% Pure active Recombinant human UbcH1.
Among ubiquitin-conjugating enzymes, the mammalian ubiquitin conjugating enzyme UbcH1, is unique in its ability to catalyze the in vitro synthesis of unanchored Lys48-linked poly-ubiquitin chains from mono- or poly-ubiquitin, E1, and ATP. In addition, UbcH1 can catalyze the cyclization of longer poly-ubiquitin chains, including tetra- and penta-ubiquitin. Recombinant UbcH1 charges and supports ubiquitinylation in vitro. Typical enzyme concentration to support conjugation in vitro is 100 nM to 1 µM.