Would you like to log in again?
TGF-beta2, human recombinant, BioVision
Supplier: BioVision
4340-50 4340-1000 4340-5
10006-578 10006-574 10006-576
TGF-beta2, human recombinant, BioVision
>98% Pure active Recombinant human TGF-beta2.
The ED₅₀ as determined by the ability of TGF-β2 to inhibit the mouse IL-4-dependent proliferation of mouse HT-2 cells is <0.2 ng/ml, corresponding to a specific activity of > 5 x 10⁶ units/mg.
TGF-β2 (Transforming Growth Factor-β2) is a member of the superfamily of homologous, disulfide-linked, homodimeric proteins that regulate the proliferation and differentiation of normal and transformed cells. Human TGF-β2 is a 25 kDa protein with each subunit containing 112 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty