Would you like to log in again?
IL-22, murine recombinant, BioVision
Supplier: BioVision
4602-1000 4602-50 4602-10
10006-890 10006-892 10006-888
IL-22, murine recombinant, BioVision
>97% Pure active Recombinant murine IL-22.
Recombinant murine IL-22 is fully biologically active when compared to a standard. The ED₅₀ as determined by its ability to induce IL-10 secretion in Colo205 cells is less than 0.5 ng/ml, corresponding to a specific activity of 2 x 10⁶ IU/mg.
Murine Interleukin-22 (IL-22), also known as T cell-derived inducible factor (ILTIF), is a cytokine that is structurally related to IL-10. Studies have shown that IL-22 induces acute-phase reactants in vitro and in vivo, suggesting that the cytokine contributes to the regulation of inflammatory responses. Recombinant murine IL-22 contains 2 x 146 amino acid chains having a total molecular mass of 33.4 kDa.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty