Human Recombinant TGF alpha (from E. coli)
Supplier: BioVision
4339-1000 4339-100 4339-20
10006-282EA 1290.08 USD
10006-282 10006-280 10006-572
Human Recombinant TGF alpha (from E. coli)
>98% Pure active Recombinant human TGF-alpha, which stimulates the proliferation of a wide range of epidermal and epithelial cells. .
The ED₅₀ as determined by the dose-dependent stimulation of proliferation of Balb-c 3T3 cells is <0.2 ng/ml, corresponding to a specific activity of >5 x 10⁶ units/mg.
Transforming Growth Factor-α (TGF-α) stimulates the proliferation of a wide range of epidermal and epithelial cells. TGF-α has been shown to play a role in the autocrine growth of certain transformed cells. Human TGF-α is a 5.5 kDa protein containing 50 amino acid residues.