Human Recombinant TDGF1 (from Cells)
Supplier: BioVision
4335-50 4335-10 4335-1000
10006-276EA 9923.7 USD
10006-276 10006-570 10006-278
Human Recombinant TDGF1 (from Cells)
>95% Pure active Recombinant human TDGF1.
Measured by its binding ability in a functional ELISA Immobilized human TDGF1 at 2 μg/ml ( 100 μl/well) can bind human ALK4 with a linear range of 0.0068 - 0.16 μg/ml.
Teratocarcinoma-derived growth factor 1, also known as epidermal growth factor-like cripto protein CR1, CRGF, and TDGF1, is a cell membrane which contains one EGF-like domain. In humans, TDGF1 is highly expressed in germ cell tumors and in colon and mammary carcinomas. TDGF1 is a member of the epidermal growth factor-cripto FRL1 cryptic protein family and is involved in the activation of several different signaling pathways during embryonic development and cellular transformation. TDGF1 regulates the activation of several signaling pathways and controls cellular transformation in embryonic status. Patients with high TDGF1 expression were statistically susceptible to a recurrence of the disease, and showed poorer disease-free survival than those with low expression. TDGF1 is a predictive marker for metachronous metastasis in colorectal cancer (CRC) patients. It is preferentially expressed in gastric and colorectal carcinomas than in their normal counterparts. TDGF1 plays a role in the determination of the epiblastic cells that subsequently give rise to the mesoderm.