Would you like to log in again?
SF20, mouse recombinant, BioVision
Supplier: BioVision
4334-10 4334-1000
10006-566 10006-568
SF20, mouse recombinant, BioVision
>97% Pure active Recombinant mouse SF20.
Mouse SF20 is a bone marrow stroma-derived growth factor. SF20 is expressed in the bone marrow, spleen stroma cells, resting mononuclear cells, resting CD8+ and CD19+ cells and activated CD8+ T cells. SF20 has been shown to bind to the surface of cells expressing the receptor TSA-1 (Thymic shared Ag-1). Among SF20’s biological activities is stimulation of the proliferation of FDCP2 cells (a mouse factor-dependent hemopoietic cell line) and mouse lymphoid cells. Chromosome 19 Open Reading Frame 10 Mouse Recombinant produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 143 amino acids and having a molecular mass of 15.8kDa. The Mouse SF20 is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty