Mouse Recombinant SCF (from E. coli)
Supplier: BioVision
4328-10 4328-50 4328-1000
10006-558EA 606.93 USD
10006-558 10006-556 10006-554
Mouse Recombinant SCF (from E. coli)
>98% Pure active Recombinant murine SCF, a hematopoietic growth factor.
The ED₅₀ as determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is <10 ng/ml, corresponding to a specific activity of >1 x 10⁵ units/mg.
Stem Cell Factor (SCF) is a hematopoietic growth factor that exerts its activity at the early stages of hematopoiesis. SCF stimulates the proliferation of myeloid, erythroid, and lymphoid progenitors in bone marrow cultures. Recombinant murine SCF is an 18.3 kDa protein containing 164 amino acid residues.