Would you like to log in again?
Adipophilin, human, recombinant, BioVision
Supplier: BioVision
4597-1000 4597-10
10006-880EA 6983.4 USD InStock
10006-880 10006-878
Adipophilin, human, recombinant, BioVision
Proteins and Peptides
>95% Pure Recombinant human Adipophilin.
Adipophilin is a 49 kDa protein that belongs to the PAT family (perilipin, adipophilin, TIP47, S3-12 and OXPAT), which comprises proteins involved in the coating of lipid droplets. ADFP is expressed in lipid droplets of vitamin A-storing hepatic stellate cells and additionally in lipid droplets of steatotic hepatocytes. ADFP expression has a role in clear cell renal carcinoma differentiation. Most recently, it has been discovered that patients with kidney cancers had high levels of ADFP in their urine, suggesting that ADFP could be an excellent marker for kidney cancer.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty