Mouse Recombinant RANTES (from E. coli)
Supplier: BioVision
4322-1000 4322-20
10006-540EA 6298.62 USD
10006-540 10006-542
Mouse Recombinant RANTES (from E. coli)
>98% Pure active Recombinant murine RANTES.
Determined by its ability to chemoattract murine T cell population using a concentration range of 1-10 ng/ml.
RANTES is a protein that has been shown to be a chemoattractant for peripheral blood monocytes. It appears to selectively attract T cells of the CD4+/CD45RO+ phenotype in vitro. Murine RANTES is a 7.8 kDa protein containing 68 amino acid residues.