Would you like to log in again?
FGF-20, human recombinant, BioVision
Supplier: BioVision
4589-100 4589-1000 4589-20
10006-872EA 715.85 USD InStock
10006-872 10006-874 10006-876
FGF-20, human recombinant, BioVision
Proteins and Peptides
>90% Pure active Recombinant human FGF-20.
The ED₅₀ as determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF-receptors is <10 ng/ml, corresponding to a specific activity of ≥1 x 10E5 units/mg.
Fibroblast Growth Factor-20 (FGF-20) is a secreted heparin binding growth factor belonging to the FGF family. FGF-20 signals through the FGFR 2c and 3c and is expressed during limb and brian development. Recombinant human FGF-20 is a 23.2 kDa protein containing 209 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty