Human Recombinant RANTES (from E. coli)
Supplier: BioVision
4321-20 4321-1000 4321-100
10006-536EA 4845.1 USD
10006-536 10006-534 10006-538
Human Recombinant RANTES (from E. coli)
>98% Pure active Recombinant human RANTES.
Determined by its ability to chemoattract human blood monocytes using a concentration range of 1-10 ng/ml.
RANTES is a CC-chemokine that can signal through the CCR1, CCR3, CCR5 and US28 (cytomegalovirus receptor) receptors. It is a chemoattractant towards monocytes, memory T cells (CD4+/CD45RO), basophils, and eosinophils. RANTES also has the capability to inhibit certain strains of HIV-1, HIV-2 and simian immunodeficiency virus (SIV). Recombinant human RANTES is a 7.8 kDa protein containing 68 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.