Would you like to log in again?
Leptin Receptor, human recombinant, BioVision
Supplier: BioVision
4582-1000 4582-50 4582-10
10006-868 10006-870 10006-810
Leptin Receptor, human recombinant, BioVision
>95% Pure active Recombinant human Leptin Receptor .
Measured by its binding ability in a functional ELISA Immobilized human Leptin at 5 μg/ml (100 μl/well) can bind human Leptin receptor with a linear range of 0.032 - 4.0 μg/ml.
Leptin receptor (LEPR), also known as OB-R and B219, is a single transmembrane-domain receptor of the cytokine receptor family. Leptin receptor exists as homodimer and binds Leptin with high affinity, thus mediates the biological function of the adipocyte-specific hormone Leptin. LEPR is expressed at high levels in hematopoietic stem cells, lymphohematopoietic cell lines, as well as adult reproductive organs. Several isoforms of LEPR have been identified, and LEPR structurally contains two hemopoietin receptor domains, a fibronectin type III domain and a WSXWS domain within the extracellular region. Interaction of leptin and leptin receptor is crucial for bodyweight and bone mass regulation in mammals through hypothalamic effects on satiety and energy expenditure. Meanwhile, research data supports a leptin receptor activation model based on ligand-induced conformational changes.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty