Human Recombinant Rank Srankl (from E. coli)
Supplier: BioVision
4318-10 4318-50 4318-1000
10006-528EA 227.66 USD
10006-528 10006-532 10006-530
Human Recombinant Rank Srankl (from E. coli)
>98% Pure active Recombinant human RANK (sRANKL), a dentritic cell membrane protein.
The ED₅₀ as determined by the dose-dependent stimulation of IL-8 production by human PBMC is <10 ng/ml.
Human RANK (receptor activator of NF-kB) is a member of the TNFR family, identified as a dentritic cell membrane protein. Human soluble RANK Ligand is a soluble 20.0 kDa polypeptide, comprising the TNF homologous region of RANKL (176 amino acid residues).