Would you like to log in again?
BMP-2, human recombinant, BioVision
Supplier: BioVision
4577-50 4577-1000 4577-10
10006-300EA 566.5 USD InStock
10006-300 10006-802 10006-800
BMP-2, human recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant human BMP-2.
The ED₅₀ as determined by the dose-dependent cytolysis of MC3T3-E1 cells is < 50 ng/ml.
BMPs (bone morphogenetic proteins) belong to the TGF-β superfamily of structurally related signaling proteins. As implied by their name, BMPs promote and regulate bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. In addition to its osteogenic activity, BMP-2 also plays an important role in cardiac morphogenesis. BMP-2 is expressed in a variety of tissues such as lung, spleen, brain, liver, prostate ovary, and small intestine. Recombinant human BMP-2 is a 26 kDa homodimeric protein consisting of two 115 amino acid polypeptide chains
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty