Human Recombinant BMP3 (from E. coli)
Supplier: BioVision
4573-1000 4573-50 4573-10
10006-788EA 93.39 USD
10006-788 10006-792 10006-790
Human Recombinant BMP3 (from E. coli)
>98% Pure active Recombinant human BMP-3.
Testing is in progress.
BMPs (bone morphogenetic proteins) belong to the TGF-β superfamily of structurally related signaling proteins. As implied by their name, BMPs promote and regulate bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. Recombinant human BMP-3 is a 31.6 kDa homodimeric protein consisting of two 134 amino acid polypeptide chains.