Would you like to log in again?
NGF-beta, human recombinant, BioVision
Supplier: BioVision
4303-1000 4303-20
10006-500EA 3759.5 USD InStock
10006-500 10006-502
NGF-beta, human recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant human NGF-beta, a potent neurotrophic factor that supports the growth and survivability of nerve and/or glial cells.
The ED₅₀ as determined using PC12 cell line is 1.2-2 ng/ml and using DRG cell line is 0.8-1.5 ng/ml.
Nerve growth factor-β (NGF-β) is a potent neurotrophic factor that supports the growth and survivability of nerve and/or glial cells. The active form of human NGF-β is a dimer, formed by two identical 119 amino acid subunits, which is held together by strong hydrophobic interactions. Recombinant human BGF-b produced in CHO cells is a homodimer, glycosylated, polypeptide chain containing 2 identical 119 amino acids and having a molecular weight of 16.95 kDa. The protein is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty