Would you like to log in again?
Caspase-2 Inhibitor Drug Screening Kit, BioVision
Supplier: BioVision
K152-100
10013-608EA 458.35 USD InStock
10013-608
Caspase-2 Inhibitor Drug Screening Kit, BioVision
Assays Protein Assays
  • Fluorescence (Ex/Em 400/505)
  • Mammalian
  • 100 assays
  • Effective means for screening caspase inhibitors using fluorometric methods
Caspases have been shown to play a crucial role in apoptosis induced by various deleterious and physiologic stimuli. Inhibition of caspases can delay apoptosis, implicating a potential role in drug screening efforts. The Caspase-2 Inhibitor Drug Screening Kit provides an effective means for screening caspase inhibitors using fluorometric methods. The assay utilizes synthetic peptide substrate VDVAD-AFC (AFC, 7-amino-4-trifluoromethyl coumarin). Active caspase-2 cleaves the synthetic substrate to release free AFC which can then be quantified by fluorometry. Compounds to be screened can directly be added to the reaction and the level of inhibition of caspase-2 activity can be determined by comparison of the fluorescence intensity in samples with and without the testing inhibitors.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty