Would you like to log in again?
Cathepsin G Activity Assay Kit, BioVision
Supplier: BioVision
K146-100
10013-602EA 478.95 USD InStock
10013-602
Cathepsin G Activity Assay Kit, BioVision
Assays Protein Assays
  • Absorbance (405 nm)
  • Mammalian
  • 100 assays
  • Detects alternative proteolytic enzymes such as the lysosomal cathepsin G proteases that can initiate or propagate proapoptotic signals
Cathepsin G is an enzymatic protein belonging to the peptidase or protease families. The protein is found in azurophil granules of neutrophilic polymorphonuclear leukocytes. The encoded protease has a specificity similar to that of chymotrypsin C, and may participate in the killing and digestion of engulfed pathogens, and in connective tissue remodeling at sites of inflammation. In BioVision’s Cathepasin G Activity assay Kit, Cathepsin G will cleave the substrate and release the dye group, pNA (4-Nitroaniline), which can be detect at 405 nm. In presence of the Cathepsin G specific inhibitor, the cleavage will be stopped. The kit provides a rapid, simple, sensitive, and reliable test suitable for the activity of Cathepsin G.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty